Sodium tetrathionate dihydrate

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Disodium tetrathionate dihydrate

461920250 461921000
ACRO461920250EA 102 EUR
ACRO461920250 ACRO461921000
Sodium tetrathionate dihydrate
Sodium tetrathionate dihydrate
Formula: Na₂O₆S₄·2H₂O
Storage Temperature: Refrigerator
MDL Number: MFCD00150868
CAS Number: 13721-29-4

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Solubility colorless to light yellow
Titration after Ion exchange ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR