2,2'-Biimidazole 97,5 HPLC_ASSAY_METHOD

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 2,2'-Diimidazole, 2,2'-Bisimidazole, 1H,1'H-2,2'-Biimidazole

465820010 465820050
ACRO465820010EA 98 EUR
ACRO465820010 ACRO465820050
2,2'-Biimidazole 97,5 HPLC_ASSAY_METHOD
2,2'-Biimidazole
Formula: C₆H₆N₄
MW: 134.14 g/mol
Melting Pt: >350°C
Density: 1,37
Storage Temperature: Ambient
MDL Number: MFCD00047014
CAS Number: 492-98-8
EINECS: 207-768-8
UN: 3265
ADR: 8 II

Order Now

Specification Test Results

Appearance Light yellow to brown to grey crystalline powder
Infrared spectrum Conforms
HPLC >=97.5%
Titration with HClO4 97.5 to 102.5%
Water =<2%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR