Lithium standard solution, 1000 mg/L Li in 2% hydrochloric acid standard for ICP
Supplier: CPACHEM
C030.2CP.L1 C030.2CP.L25 C030.2CP.L5
ACSDC030.2CP.L1EA 84.9 EUR
ACSDC030.2CP.L1 ACSDC030.2CP.L25 ACSDC030.2CP.L5
Lithium standard solution, 1000 mg/L Li in 2% hydrochloric acid standard for ICP
Standards Cationic Standards Lithium Standards