Lanthanum standard solution, 10000 mg/L La in 5% nitric acid standard for ICP
Supplier: CPACHEM
C129.5NP.L1 C129.5NP.L25 C129.5NP.L5
ACSDC129.5NP.L1EA 192 EUR
ACSDC129.5NP.L1 ACSDC129.5NP.L25 ACSDC129.5NP.L5
Lanthanum standard solution, 10000 mg/L La in 5% nitric acid standard for ICP
Standards Cationic Standards Lanthanum Standards