Zinc Standard Solution, 10000 mg/L Zn in dil. Nitric Acid Standard for ICP-MS
Supplier: ACTIVE MOTIF
M169.5NP.L1
ACSDM169.5NP.L1EA 238 EUR
ACSDM169.5NP.L1
Zinc Standard Solution, 10000 mg/L Zn in dil. Nitric Acid Standard for ICP-MS
Standards Cationic Standards Zinc Standards
Mg 10000 mg/L in diluted nitric acid for ICP/MS.
Shelf life on ship date: 36 months - store under normal laboratory conditions, at temperatures between 15 to 25 °C.
Certifications: ISO 17034, ISO 17025, ISO 9001.