Bioanalyzer Small RNA Analysis
Supplier: AGILENT
5067-1548 5067-1549 5067-1550
AGLS5067-1548EA 1950 EUR
AGLS5067-1548 AGLS5067-1549 AGLS5067-1550
Bioanalyzer Small RNA Analysis
Electrophoresis Systems Capillary Electrophoresis Systems
High resolution small RNA electrophoresis of fragments from 6 to 150 nt.
  • High resolution analysis, with a sizing range from 6 to 150 nucleotide, allows for separation of miRNA smears from tRNA
  • Minimal sample consumption, only use as little as 50 pg of purified miRNA or 10 ng of total RNA for analysis in 1 μl volume
  • A faster and more sensitive alternative to agarose and polyacrylamide gels analysis for detection of small RNA
  • Compatibility with multiple sample types enables analysis of total RNA, fractionated, or synthetic samples