Brilliant probe multiplex MM
Supplier: AGILENT
600553
AGLS600553EA 408 EUR
AGLS600553
Brilliant probe multiplex MM
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
The brilliant multiplex QPCR master mix allows you to amplify up to four targets in a single real-time PCR Reaction.
  • Maximises analysis of limited or rare samples
  • Allows detection of multiple targets plus an internal control
  • More economical per sample than singleplex, while saving time and increasing throughput
It provides sufficient reaction components to accurately quantify both low-and-high-abundance targets in the same tube.
The sensitivity remains equivalent to that seen in singleplex.