Large Fragment Analysis Kits
Supplier: AGILENT
DNF-464-0500 DNF-492-0500 DNF-492-1000
AGLSDNF-464-0500EA 1750 EUR
AGLSDNF-464-0500 AGLSDNF-492-0500 AGLSDNF-492-1000
Large Fragment Analysis Kits
Electrophoresis Systems Capillary Electrophoresis Systems
DNA analysis for large smears, fragment sizing, long read NGS library QC and single molecule sequencing.
  • Broad concentration range - Each kit has different input concentration ranges specific for DNA fragments and smears
  • Reliable sizing - Kits for large sized DNA fragments and long-read NGS libraries help ensure accurate and precise sizing up to 50 kb
  • Low sample volume - Only 2 µl of sample required minimising sample loss to QC steps
The large fragment kits for the fragment analyser systems are used for the automated quality and quantity analysis ofMore Product Information