Femto Pulse System
Supplier: AGILENT
DNF-325-0010 DNF-325-0075 FP-OQ GP-435-0100 M5330AA M5330AA-001
AGLSDNF-325-0010EA 28.3 EUR
AGLSDNF-325-0010 AGLSDNF-325-0075 AGLSFP-OQ AGLSGP-435-0100 AGLSM5330AA AGLSM5330AA-001
Femto Pulse System
Electrophoresis Systems Capillary Electrophoresis Systems
Pulsed-field capillary electrophoresis system for QC of high molecular weight DNA and RNA samples.
  • Large fragment resolution - separate high molecular weight DNA smears and fragments through 165 kb in about 1,5 hours
  • Femtogram level sensitivity - achieve 10 times higher sensitivity for nucleic acid smears and up to 100 times higher for nucleic acid fragments
  • Simple sample preparation - minimise sample degradation with fewer pipetting steps and stable reagents
  • Powerful analysis software - one intuitive software program automates analysis of diverse sample