Thermometer, stirring, mercury filled, measuring range:-10:110/1C, length:260mm, diameter:6-7mm, solid stem, white backed, durable pigment. Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclusively intended to 1 * 1 items
Supplier: Amarell
Thermometer, stirring, mercury filled, measuring range:-10:110/1C, length:260mm, diameter:6-7mm, solid stem, white backed, durable pigment. Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclusively intended to 1 * 1 items