Thermometer, refill, pocket, red special liquid filled, measuring range: -10:100/1C, length:125mm, diameter:9mm, colourless, withou t metal case, capillary lens front. 1 * 1 items
Supplier: Amarell
Thermometer, refill, pocket, red special liquid filled, measuring range: -10:100/1C, length:125mm, diameter:9mm, colourless, withou t metal case, capillary lens front. 1 * 1 items