Shoe covers, AlphaTec® Sure Step, model 403
Supplier: Ansell
WHSSS-00403-00
ANSEWHSSS-00403-00EA 643 EUR
ANSEWHSSS-00403-00
Shoe covers, AlphaTec® Sure Step, model 403
Disposable Garments
A disposable shoe cover that provides slip-resistance to the underside of the feet.
  • Liquid resistant material
  • Low linting
  • Anti-static
  • Elasticated opening
Weight: 65 g/m²