Anti-CD50 Mouse Monoclonal Antibody [Clone: MEM-171]

Supplier: ANTIBODIES.COM
A85863-100
ANTIA85863-100EA 260 EUR
ANTIA85863-100
Anti-CD50 Mouse Monoclonal Antibody [Clone: MEM-171]
Antibodies
Mouse monoclonal [MEM-171] antibody to CD50 for flow cytometry and IP with samples derived from human.

  • Validated applications: Flow cytometry, IP
  • Recommended dilutions: Flow cytometry: 4 µg/ml

Type: Primary
Antigen: CD50
Clonality: Monoclonal
Clone: MEM-171
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR