2′-(4-Methylumbelliferyl)-α-D-N-acetylneuraminic acid sodium salt hydrate

Supplier: Apollo Scientific
Warning

Synonyms: 4-MUNANA

BIB6114-100MG BIB6114-25MG
APOSBIB6114-100MGEA 393 EUR
APOSBIB6114-100MG APOSBIB6114-25MG
2′-(4-Methylumbelliferyl)-α-D-N-acetylneuraminic acid sodium salt hydrate
2′-(4-Methylumbelliferyl)-alpha-D-N-acetylneuraminic acid sodium salt hydrate
Formula: C₂₁H₂₄NNaO₁₁·xH₂O
Storage Temperature: Freezer
MDL Number: MFCD00057309
CAS Number: 76204-02-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR