3-O-Allyl-1,2:5,6-di-O-isopropylidene-α-D-glucofuranose ≥98%

Supplier: Apollo Scientific

Synonyms: 1,2:5,6-Di-O-isopropylidene-3-O-2-propen-1-yl-α-D-glucofuranose

BICL4319-10G BICL4319-25G BICL4319-50G
APOSBICL4319-10GEA 422 EUR
APOSBICL4319-10G APOSBICL4319-25G APOSBICL4319-50G
3-O-Allyl-1,2:5,6-di-O-isopropylidene-α-D-glucofuranose ≥98%
3-O-Allyl-1,2:5,6-di-O-isopropylidene-α-D-glucofuranose
Formula: C₁₅H₂₄O₆
MW: 300.347 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01725444
CAS Number: 20316-77-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR