Dovitinib lactate

Supplier: Apollo Scientific
Warning

Synonyms: CHIR-258, TKI-258, 4-Amino-5-fluoro-3-(6-(4-methylpiperazin-1-yl)-1H-benzo[d]imidazol-2-yl)quinolin-2(1H)-one 2-hydroxypropanoate

BIFK0016-5MG
APOSBIFK0016-5MGEA 69.6 EUR
APOSBIFK0016-5MG
Dovitinib lactate
Dovitinib lactate

Lactate Salt, >99%

Formula: C₂₁H₂₁FN₆O·C₃H₆O₃
Storage Temperature: Freezer
MDL Number: MFCD16038905
CAS Number: 692737-80-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR