Quercetin dihydrate

Supplier: Apollo Scientific

Synonyms: 2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-chromen-4-one dihydrate

BIK9012-25G BIK9012-100G
APOSBIK9012-25GEA 87 EUR
APOSBIK9012-25G APOSBIK9012-100G
Quercetin dihydrate
Quercetin dihydrate
Formula: C₁₅H₁₄O₉
Melting Pt: 315 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149487
CAS Number: 6151-25-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR