Spermine-(N,N'-bis(3-aminopropyl)-1,4-butanediamine) ≥98%

Supplier: Apollo Scientific
Danger

Synonyms: N1,N1'-(Butane-1,4-diyl)bis(propane-1,3-diamine), Spermine

BIS2403-25G BIS2403-5G
APOSBIS2403-25GEA 270 EUR
APOSBIS2403-25G APOSBIS2403-5G
Spermine-(N,N'-bis(3-aminopropyl)-1,4-butanediamine) ≥98%
Spermine (N,N'-Bis(3-aminopropyl)-1,4-butanediamine)
Formula: C₁₀H₂₆N₄
Boiling Pt: 150 °C/5mmHg
Melting Pt: 28…30 °C
Density: 0.937 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008215
CAS Number: 71-44-3
EINECS: 200-754-2
UN: 3259
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR