Saponin (from Quillaja Bark)

Supplier: Apollo Scientific
Warning

BIS4332-1KG
APOSBIS4332-1KGEA 1560 EUR
APOSBIS4332-1KG
Saponin (from Quillaja Bark)
Saponin

Surfactant which allows penetration of proteins throughcell membranes

Melting Pt: 158 °C (dec.)
Density: 0.5 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00081981
CAS Number: 8047-15-2
EINECS: 232-462-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR