Spermine-(N,N'-bis(3-aminopropyl)-1,4-butanediamine)dihydrate

Supplier: Apollo Scientific
Danger

Synonyms: Spermine Dihydrate

BIS5944-25G BIS5944-5G
APOSBIS5944-25GEA 418 EUR
APOSBIS5944-25G APOSBIS5944-5G
Spermine-(N,N'-bis(3-aminopropyl)-1,4-butanediamine)dihydrate
Spermine (N,N'-Bis(3-aminopropyl)-1,4-butanediamine) dihydrate
Formula: C₁₀H₂₆N₄·2H₂O
Melting Pt: 63…64 °C
Storage Temperature: Refrigerator
CAS Number: 403982-64-9
UN: 3259
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR