N,N,N',N'-Tetramethylethylenediamine (TEMED)

Supplier: Apollo Scientific
Danger

Synonyms: 1,2-Bis(dimethylamino)ethane, TMEDA, N,N,N',N'-Tetramethylethylenediamine

BIT7140-100ML BIT7140-250ML
APOSBIT7140-100MLEA 29 EUR
APOSBIT7140-100ML APOSBIT7140-250ML
N,N,N',N'-Tetramethylethylenediamine (TEMED)
N,N,N',N'-Tetramethylethylenediamine (TEMED)
Formula: C₆H₁₆N₂
Boiling Pt: 120…122 °C
Melting Pt: –55 °C
Density: 0.775 g/cm³ (25 °C)
Flash Pt: 20 °C
Storage Temperature: Ambient
MDL Number: MFCD00008335
CAS Number: 110-18-9
UN: 2372
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR