Ammonium tetrathiomolybdate

Supplier: Apollo Scientific
Warning

IN1087-10G IN1087-50G IN1087-1G
APOSIN1087-10GEA 306 EUR
APOSIN1087-10G APOSIN1087-50G APOSIN1087-1G
Ammonium tetrathiomolybdate
Ammonium tetrathiomolybdate
Formula: H₁₀MoN₂S₄
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00136013
CAS Number: 15060-55-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR