Antimony(III) chloride 99,9%

Supplier: Apollo Scientific
Danger

Synonyms: Antimony trichloride

IN1108-100G IN1108-25G
APOSIN1108-100GEA 304 EUR
APOSIN1108-100G APOSIN1108-25G
Antimony(III) chloride 99,9%
Antimony (III) chloride
Formula: SbCl₃
Boiling Pt: 283 °C
Melting Pt: 73.4 °C
Density: 3.14 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011212
CAS Number: 10025-91-9
EINECS: 233-047-2
UN: 1733
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR