Gallium trinitrate hydrate 99,9%

Supplier: Apollo Scientific
Warning

IN1885-25G
APOSIN1885-25GEA 306 EUR
APOSIN1885-25G
Gallium trinitrate hydrate 99,9%
Gallium trinitrate hydrate
Formula: GaN₃O₉·H₂O
MW: 273.75 g/mol
Melting Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00149723
CAS Number: 69365-72-6
EINECS: 236-815-5
UN: 1477
ADR: 5.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR