Rhenium dioxide 99,9%

Supplier: Apollo Scientific

Synonyms: Rhenium (IV) oxide, Rhenium(iv)oxide

IN2995-1G IN2995-5G
APOSIN2995-1GEA 125 EUR
APOSIN2995-1G APOSIN2995-5G
Rhenium dioxide 99,9%
Rhenium dioxide
Formula: O₂Re
Melting Pt: 1000 °C
Density: 11.4 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016305
CAS Number: 12036-09-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR