Samarium(III) oxide 99.999%

Supplier: Apollo Scientific
Warning

IN3100-100G
APOSIN3100-100GEA 574 EUR
APOSIN3100-100G
Samarium(III) oxide 99.999%
Samarium (III) oxide
Formula: O₃Sm₂
Melting Pt: 2300 °C
Density: 8.347 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011237
CAS Number: 12060-58-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR