Silver(I) oxide 99,5%

Supplier: Apollo Scientific
Danger

Synonyms: di-Silver oxide, Disilver oxide, Silver oxide

IN3238-1G
APOSIN3238-1GEA 75.4 EUR
APOSIN3238-1G
Silver(I) oxide 99,5%
Silver(I) oxide
Formula: Ag₂O
Melting Pt: 230 °C (dec.)
Density: 7.2 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003404
CAS Number: 20667-12-3
EINECS: 243-957-1
UN: 1479
ADR: 5.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR