Titanium(IV) oxide rutile 99.99%

Supplier: Apollo Scientific
Warning

Synonyms: Pigment White 6, Rutile, Titanium dioxide rutile

IN3706-100G IN3706-25G IN3706-250G
APOSIN3706-100GEA 219 EUR
APOSIN3706-100G APOSIN3706-25G APOSIN3706-250G
Titanium(IV) oxide rutile 99.99%
Titanium (IV) oxide rutile
Formula: O₂Ti
Boiling Pt: 2500…3000 °C
Melting Pt: 1855 °C
Density: 4.26 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011269
CAS Number: 1317-80-2
EINECS: 215-282-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR