Tungsten trioxide 99+%

Supplier: Apollo Scientific
Warning

Synonyms: Tungsten (VI) oxide

IN3736-100G IN3736-500G
APOSIN3736-100GEA 161 EUR
APOSIN3736-100G APOSIN3736-500G
Tungsten trioxide 99+%
Tungsten trioxide
Formula: O₃W
Melting Pt: 1473 °C
Density: 7.16 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011466
CAS Number: 1314-35-8
EINECS: 215-231-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR