4-Iodo-o-toluidine

Supplier: Apollo Scientific
Warning

Synonyms: 2-Amino-5-iodotoluene, 4-Iodo-2-methylaniline

OR0552-100G OR0552-25G OR0552-5G
APOSOR0552-100GEA 95.7 EUR
APOSOR0552-100G APOSOR0552-25G APOSOR0552-5G
4-Iodo-o-toluidine
4-Iodo-o-toluidine
Formula: C₇H₈IN
Melting Pt: 86…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00025299
CAS Number: 13194-68-8
EINECS: 236-154-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR