2,3-Dimethylbenzonitrile 98%

Supplier: Apollo Scientific

OR0561-25G
APOSOR0561-25GEA 82.8 EUR
APOSOR0561-25G
2,3-Dimethylbenzonitrile 98%
2,3-Dimethylbenzonitrile
Formula: C₉H₉N
Boiling Pt: 64…66 °C/0.8mmHg
Melting Pt: 23…25 °C
Storage Temperature: Ambient
MDL Number: MFCD00019744
CAS Number: 5724-56-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR