3-Aminopiperidine hydrochloride, N1-BOC protected

Supplier: Apollo Scientific
Warning

Synonyms: tert-Butyl 3-aminopiperidine-1-carboxylate hydrochloride, 3-Amino-1-(tert-butoxycarbonyl)piperidine hydrochloride

OR0735-10G OR0735-1G OR0735-5G
APOSOR0735-10GEA 487 EUR
APOSOR0735-10G APOSOR0735-1G APOSOR0735-5G
3-Aminopiperidine hydrochloride, N1-BOC protected
3-Aminopiperidine hydrochloride, N1-BOC protected
Formula: C₁₀H₂₀N₂O₂·ClH
Melting Pt: 181…182 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD06691424
CAS Number: 1000796-62-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR