6-(Methylsulphonyl)oxindole 97%
Supplier: Apollo Scientific
Warning
OR10557-250MG
APOSOR10557-250MGEA 45.1 EUR
APOSOR10557-250MG
6-(Methylsulphonyl)oxindole 97%
6-(Methylsulphonyl)oxindole
Formula: C₉H₉NO₃S
Melting Pt: 218…220 °C
Storage Temperature: Ambient
MDL Number: MFCD02179607
CAS Number: 850429-63-9