2,3-Dibromopropyl acrylate

Supplier: Apollo Scientific
Warning

Synonyms: Acrylic Acid 2,3-Dibromopropyl Ester

OR1062-5G OR1062-1G OR1062-25G
APOSOR1062-5GEA 34.8 EUR
APOSOR1062-5G APOSOR1062-1G APOSOR1062-25G
2,3-Dibromopropyl acrylate
2,3-Dibromopropyl acrylate
Formula: C₆H₈Br₂O₂
Boiling Pt: 112 °C/1.2mmHg
Storage Temperature: Ambient
MDL Number: MFCD00059461
CAS Number: 19660-16-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR