2-Bromo-1-(2,3-dihydro-1-benzofuran-5-yl)ethanone

Supplier: Apollo Scientific
Danger

Synonyms: 2-Bromo-1-(2,3-dihydrobenzofuran-5-yl)ethanone

OR111097-1G OR111097-250MG
APOSOR111097-1GEA 256 EUR
APOSOR111097-1G APOSOR111097-250MG
2-Bromo-1-(2,3-dihydro-1-benzofuran-5-yl)ethanone
2-Bromo-1-(2,3-dihydro-1-benzofuran-5-yl)ethanone
Formula: C₁₀H₉BrO₂
Storage Temperature: Ambient
MDL Number: MFCD08444468
CAS Number: 151427-19-9
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR