3-(4-Bromophenyl)-5-methyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone

Supplier: Apollo Scientific

OR12047-1G OR12047-5G
APOSOR12047-1GEA 180 EUR
APOSOR12047-1G APOSOR12047-5G
3-(4-Bromophenyl)-5-methyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone
3-(4-Bromophenyl)-5-methyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone
Formula: C₁₅H₁₁BrN₂OS
Melting Pt: 344…346 °C
Storage Temperature: Ambient
MDL Number: MFCD08689700
CAS Number: 937601-63-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR