Lithium 2-[(4-methylpiperazin-1-yl)methyl]benzoate 90%

Supplier: Apollo Scientific
Warning

Synonyms: [(4-Methylpiperazin-1-yl)methyl]benzoic acid lithium salt

OR12266-1G OR12266-250MG
APOSOR12266-1GEA 371 EUR
APOSOR12266-1G APOSOR12266-250MG
Lithium 2-[(4-methylpiperazin-1-yl)methyl]benzoate 90%
Lithium 2-[(4-methylpiperazin-1-yl)methyl]benzoate
Formula: C₁₃H₁₇N₂O₂·Li
Melting Pt: 337…348.5 °C
Storage Temperature: Ambient
MDL Number: MFCD09064954
CAS Number: 915707-44-7
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR