1,1-Dimethyl-2-morpholin-4-yl-ethylamine 97%

Supplier: Apollo Scientific
Danger

Synonyms: 4-(2-Amino-2-methylprop-1-yl)morpholine

OR12326-1G OR12326-250MG
APOSOR12326-1GEA 237 EUR
APOSOR12326-1G APOSOR12326-250MG
1,1-Dimethyl-2-morpholin-4-yl-ethylamine 97%
1,1-Dimethyl-2-morpholin-4-ylethylamine
Formula: C₈H₁₈N₂O
Storage Temperature: Ambient
MDL Number: MFCD01714129
CAS Number: 6105-75-5
UN: 2735
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR