tert-Butyl-3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate 98%

Supplier: Apollo Scientific
Danger

Synonyms: tert-Butyl 3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate, N-Boc-Nortropinone

OR12337-25G OR12337-100G OR12337-5G
APOSOR12337-25GEA 37.7 EUR
APOSOR12337-25G APOSOR12337-100G APOSOR12337-5G
tert-Butyl-3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate 98%
tert-Butyl-3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate
Formula: C₁₂H₁₉NO₃
Melting Pt: 70…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00673779
CAS Number: 185099-67-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR