4-Hydroxy-2-quinolinecarboxylic acid hydrate

Supplier: Apollo Scientific
Warning

Synonyms: 4-Hydroxyquinaldic acid hydrate, Kynurenic acid hydrate

OR12784-1G OR12784-5G
APOSOR12784-1GEA 23.2 EUR
APOSOR12784-1G APOSOR12784-5G
4-Hydroxy-2-quinolinecarboxylic acid hydrate
4-Hydroxy-2-quinolinecarboxylic acid hydrate
Formula: C₁₀H₇NO₃·xH₂O
Melting Pt: 275 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006753
CAS Number: 345909-35-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR