2-(4-Methoxyphenoxy)-1-ethanol

Supplier: Apollo Scientific
Warning

Synonyms: 2-(4-Methoxyphenoxy)ethan-1-ol, 4-(2-Hydroxyethoxy)anisole

OR14833-1G
APOSOR14833-1GEA 0 EUR
APOSOR14833-1G
2-(4-Methoxyphenoxy)-1-ethanol
2-(4-Methoxyphenoxy)-1-ethanol
Formula: C₉H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00016568
CAS Number: 5394-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR