2-(5-Methyl-2-phenyl-1,3-thiazol-4-yl)acetic acid

Supplier: Apollo Scientific
Warning

OR15233-1G OR15233-5G
APOSOR15233-1GEA 232 EUR
APOSOR15233-1G APOSOR15233-5G
2-(5-Methyl-2-phenyl-1,3-thiazol-4-yl)acetic acid
2-(5-Methyl-2-phenyl-1,3-thiazol-4-yl)acetic acid
Formula: C₁₂H₁₁NO₂S
Melting Pt: 135…137 °C
Storage Temperature: Ambient
MDL Number: MFCD00277218
CAS Number: 101736-22-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR