(1S,2R)-N1-(tert-Butoxycarbonyl)-1,2-cyclohexanediamine

Supplier: Apollo Scientific
Warning

Synonyms: (1S,2R)-N1-Boc-1,2-cyclohexanediamine, (1S,2R)-2-Amino-1-(tert-butoxycarbonylamino)cyclohexane, tert-Butyl ((1S,2R)-2-aminocyclohexyl)carbamate, (1S,2R)-2-Amino-1-(Boc-amino)cyclohexane

OR15997-1G OR15997-5G OR15997-10G
APOSOR15997-1GEA 114 EUR
APOSOR15997-1G APOSOR15997-5G APOSOR15997-10G
(1S,2R)-N1-(tert-Butoxycarbonyl)-1,2-cyclohexanediamine
(1S,2R)-N1-(tert-Butoxycarbonyl)-1,2-cyclohexanediamine
Formula: C₁₁H₂₂N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD09952105
CAS Number: 365996-30-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR