N-Methyl-1-(methylsulphonyl)piperidin-4-amine

Supplier: Apollo Scientific

Synonyms: 4-(Methylamino)-1-(methylsulphonyl)piperidine

OR16035-1G OR16035-50MG OR16035-100MG OR16035-250MG OR16035-500MG
APOSOR16035-1GEA 357 EUR
APOSOR16035-1G APOSOR16035-50MG APOSOR16035-100MG APOSOR16035-250MG APOSOR16035-500MG
N-Methyl-1-(methylsulphonyl)piperidin-4-amine
N-Methyl-1-(methylsulphonyl)piperidin-4-amine
Formula: C₇H₁₆N₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD08061933
CAS Number: 438585-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR