1-(2-Chloroethyl)-3-(7-nitro-1,1,3,3,6-pentamethylindan-5-yl)urea

Supplier: Apollo Scientific

Synonyms: 5-{[(2-Chloroethyl)carbamoyl]amino}-2,3-dihydro-7-nitro-1,1,3,3,6-pentamethyl-1H-indene

OR16158-1G
APOSOR16158-1GEA 105 EUR
APOSOR16158-1G
1-(2-Chloroethyl)-3-(7-nitro-1,1,3,3,6-pentamethylindan-5-yl)urea
1-(2-Chloroethyl)-3-(7-nitro-1,1,3,3,6-pentamethylindan-5-yl)urea
Formula: C₁₇H₂₄ClN₃O₃
Storage Temperature: Ambient
MDL Number: MFCD11109512
CAS Number: 1211016-69-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR