Bis(cyclopentadienyl)titanium dichloride

Supplier: Apollo Scientific
Danger

Synonyms: Titanocenedichloride, Bis(cyclopentadienyl)titanium(IV) dichloride, Titanocene dichloride

OR17913-10G OR17913-250G OR17913-50G
APOSOR17913-10GEA 21.9 EUR
APOSOR17913-10G APOSOR17913-250G APOSOR17913-50G
Bis(cyclopentadienyl)titanium dichloride
Bis(cyclopentadienyl)titanium dichloride
Formula: C₁₀H₁₀Cl₂Ti
Melting Pt: 260…280 °C (dec.)
Density: 1.6 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003723
CAS Number: 1271-19-8
EINECS: 215-035-9
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR