2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl 98%

Supplier: Apollo Scientific
Warning

Synonyms: racemic-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, rac-BINAP, (+/-)-(1,1'-Binaphthalene-2,2'-diyl)bis(diphenylphosphine), (±)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, (+/-)-BINAP

OR18264-25G OR18264-5G
APOSOR18264-25GEA 66.7 EUR
APOSOR18264-25G APOSOR18264-5G
2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl 98%
2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
Formula: C₄₄H₃₂P₂
Melting Pt: 283…286 °C
Storage Temperature: Ambient
MDL Number: MFCD00010805
CAS Number: 98327-87-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR