2-(Ethylthio)nicotinoyl chloride

Supplier: Apollo Scientific
Danger

OR21412-1G
APOSOR21412-1GEA 29 EUR
APOSOR21412-1G
2-(Ethylthio)nicotinoyl chloride
2-(Ethylthio)nicotinoyl chloride
Formula: C₈H₈ClNOS
Melting Pt: 53…55 °C (dec.)
Storage Temperature: Ambient
CAS Number: 123116-01-8
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR