7-Methyl-4-indanol

Supplier: Apollo Scientific
Warning

Synonyms: 7-Methyl-2,3-dihydro-1H-inden-4-ol, 7-Methylindan-4-ol

OR21818-1G OR21818-5G OR21818-10G
APOSOR21818-1GEA 116 EUR
APOSOR21818-1G APOSOR21818-5G APOSOR21818-10G
7-Methyl-4-indanol
7-Methyl-4-indanol
Formula: C₁₀H₁₂O
MDL Number: MFCD00173675
CAS Number: 16400-13-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR