4-Iodo-5-methyl-3-phenylisoxazole

Supplier: Apollo Scientific
Warning

OR22985-250MG
APOSOR22985-250MGEA 69.6 EUR
APOSOR22985-250MG
4-Iodo-5-methyl-3-phenylisoxazole
4-Iodo-5-methyl-3-phenylisoxazole
Formula: C₁₀H₈INO
Storage Temperature: Ambient
MDL Number: MFCD02681888
CAS Number: 31295-66-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR